2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%
Thermo Scientific Chemicals

2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%

CAS: 306960-77-0 | C6H3F3N2O2 | 192.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H54088.03
also known as H54088-03
Price (USD)
146.65
Special offer
Online exclusive
Ends: 30-Jun-2026
172.00
Save 25.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
146.65
Special offer
Online exclusive
Ends: 30-Jun-2026
172.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS306960-77-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or crystalline powder or powder
Assay (GC)≥96.0%
Melting point170-178°C
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only