(Boc-aminooxy)acetic acid, 98+%
(Boc-aminooxy)acetic acid, 98+%
(Boc-aminooxy)acetic acid, 98+%
Thermo Scientific Chemicals

(Boc-aminooxy)acetic acid, 98+%

CAS: 42989-85-5 | C7H13NO5 | 191.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H54109.14
also known as H54109-14
Price (USD)
819.65
Online Exclusive
910.00
Save 90.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
819.65
Online Exclusive
910.00
Save 90.35 (10%)
Each
Chemical Identifiers
CAS42989-85-5
IUPAC Name2-({[(tert-butoxy)carbonyl]amino}oxy)acetic acid
Molecular FormulaC7H13NO5
InChI KeyQBXODCKYUZNZCY-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NOCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.0%
(Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Karen L.Christman; Rebecca M.Broyer; Zachary P.Tolstyka; Heather D.Maynard. Site-specific protein immobilization through N-terminal oxime linkages. J. Mater. Chem. 2007, 17, (19),2021-2027
  2. Damien Forget; Dr. Didier Boturyn; Dr. Eric Defrancq; Prof. Jean Lhomme; Prof. Pascal Dumy. Highly efficient synthesis of peptide-oligonucleotide conjugates: Chemoselective oxime and thiazolidine formation. Chemistry - A European Journal. 2001, 7, (18),3976-3984