Specifications
CAS1006876-28-3
Chemical Name or Material2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester
Formula Weight276.14
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more