5-Fluoroindoline, 97%
5-Fluoroindoline, 97%
5-Fluoroindoline, 97%
Thermo Scientific Chemicals

5-Fluoroindoline, 97%

CAS: 2343-22-8 | C8H8FN | 137.157 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H54158.06
also known as H54158-06
Price (USD)
272.65
Special offer
Online exclusive
Ends: 28-Dec-2026
321.00
Save 48.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
272.65
Special offer
Online exclusive
Ends: 28-Dec-2026
321.00
Save 48.35
Each
Chemical Identifiers
CAS2343-22-8
IUPAC Name5-fluoro-2,3-dihydro-1H-indole
Molecular FormulaC8H8FN
InChI KeyNXQRMQIYCWFDGP-UHFFFAOYSA-N
SMILESFC1=CC2=C(NCC2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormOil
Assay (unspecified)>96.0%
Appearance (Color)Colorless to light green or yellow
5-Fluoroindoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoroindoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. David C.Tully; Hong Liu; Arnab K.Chatterjee; Phil B.Alper; Jennifer A.Williams; Michael J.Roberts; Daniel Mutnick; David H.Woodmansee; Thomas Hollenbeck; Perry Gordon; Jonathan Chang; Tove Tuntland; Christine Tumanut; Jun Li; Jennifer L.Harris; Donald S.Karanewsky. Arylaminoethyl carbamates as a novel series of potent and selective cathepsin S inhibitors. Bioorganic & Medicinal Chemistry Letters. 2006, 16, (19),5107-5111