4-Cyanobenzyl alcohol, 97%
4-Cyanobenzyl alcohol, 97%
4-Cyanobenzyl alcohol, 97%
Thermo Scientific Chemicals

4-Cyanobenzyl alcohol, 97%

CAS: 874-89-5 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54283.03
also known as H54283-03
Price (USD)
67.40
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
67.40
Each
Specifications
CAS874-89-5
Chemical Name or Material4-Cyanobenzyl alcohol
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Formula Weight133.15
Health Hazard 1H302+H312+H332
View more
4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Nungruethai Yoswathananont; Kohei Nitta;Yumi Nishiuchi; Masaaki Sato. Continuous hydrogenation reactions in a tube reactor packed with Pd/C. Chem. Commun. 2005, (1),40-42
  2. Kohsuke Mori; Kazuya Yamaguchi; Tomoo Mizugaki; Kohki Ebitani; Kiyotomi Kaneda. Catalysis of a hydroxyapatite-bound Ru complex:efficient heterogeneousoxidation of primary amines to nitriles in thepresence of molecular oxygen. Chem. Commun. 2001, (5),461-462