4-(6-Amino-3-pyridyl)-1-Boc-piperazine, 97%
4-(6-Amino-3-pyridyl)-1-Boc-piperazine, 97%
4-(6-Amino-3-pyridyl)-1-Boc-piperazine, 97%
Thermo Scientific Chemicals

4-(6-Amino-3-pyridyl)-1-Boc-piperazine, 97%

CAS: 571188-59-5 | C14H22N4O2 | 278.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54315.06
also known as H54315-06
Price (USD)
532.65
Special offer
Online exclusive
Ends: 30-Jun-2026
626.00
Save 93.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
532.65
Special offer
Online exclusive
Ends: 30-Jun-2026
626.00
Save 93.35 (15%)
Each
Specifications
Beilstein10173494
CAS571188-59-5
Chemical Name or Material4-(6-Amino-3-pyridyl)-1-Boc-piperazine
Formula Weight278.35
Health Hazard 1H315-H319-H335
View more
4-(6-Amino-3-pyridyl)-1-Boc-piperazine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(6-Amino-3-pyridyl)-1-Boc-piperazine is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only