3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H54339.03
also known as H54339-03
1 g
Catalog number H54339.03
also known as H54339-03
Price (USD)
318.65
Online Exclusive
354.00
Save 35.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
318.65
Online Exclusive
354.00
Save 35.35 (10%)
Each
Add to cart
Specifications
CAS1083326-28-6
Chemical Name or Material3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester
Formula Weight360.24
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only