Methyl pyrrole-3-carboxylate, 97%
Methyl pyrrole-3-carboxylate, 97%
Methyl pyrrole-3-carboxylate, 97%
Methyl pyrrole-3-carboxylate, 97%
Thermo Scientific Chemicals

Methyl pyrrole-3-carboxylate, 97%

CAS: 2703-17-5 | C6H7NO2 | 125.127 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54340.06
also known as H54340-06
Price (USD)
289.65
Online Exclusive
322.00
Save 32.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
289.65
Online Exclusive
322.00
Save 32.35 (10%)
Each
Add to cart
Specifications
CAS2703-17-5
Chemical Name or MaterialMethyl pyrrole-3-carboxylate
Formula Weight125.13
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hugh J.Anderson; Charles E.Loader; Ru Xun Xu; Nghia Le, Niall J.Gogan,; Robert McDonald; Louise G.Edwards. Pyrrole chemistry. XXVIII. Substitution reactions of 1-(phenylsulfonyl)pyrrole and some derivatives. Canadian Journal of Chemistry. 1985, 63, (4),896-902
  2. Brian L.Bray; Joseph M.Muchowski. α-(Dialkylamino)-α-pyrrolylacetonitriles. A potpourri of useful synthetic transformations. Canadian Journal of Chemistry. 1990, 68, (8),1305-1308