2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 96%
2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 96%
2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 96%
Thermo Scientific Chemicals

2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 96%

CAS: 944401-55-2 | C11H18BN3O2 | 235.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H54375.03
also known as H54375-03
Price (USD)
245.65
Special offer
Online exclusive
Ends: 30-Jun-2026
289.00
Save 43.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
245.65
Special offer
Online exclusive
Ends: 30-Jun-2026
289.00
Save 43.35 (15%)
Each
Specifications
CAS944401-55-2
Chemical Name or Material2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester
Formula Weight235.09
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only