Methyl hydrogen terephthalate, 99+%
Methyl hydrogen terephthalate, 99+%
Methyl hydrogen terephthalate, 99+%
Thermo Scientific Chemicals

Methyl hydrogen terephthalate, 99+%

CAS: 1679-64-7 | C9H8O4 | 180.159 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H54391.06
also known as H54391-06
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.80
Save 7.15 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.80
Save 7.15 (15%)
Each
Chemical Identifiers
CAS1679-64-7
IUPAC Name4-(methoxycarbonyl)benzoic acid
Molecular FormulaC9H8O4
InChI KeyREIDAMBAPLIATC-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>99.0%
Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

Solubility
Slightly soluble in water. Also soluble in methanol, benzene and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. James K.Hurst; Henry Taube. The reduction by metal ions of fumaratopentaammine cobalt(III) complexes. J. Am. Chem. Soc. 1968, 90,(5), 1178-1186.
  2. L.N.Owen; R.Sridhar. Cytotoxic compounds. Part X. Carbamates derived from NN-bis-(2-chloroethyl)-m- and NN-bis-(2-chloroethyl)-p-phenylenediamine. J. Chem. Soc. C. 1970, (3), 472-475.