2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 97%
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 97%
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 97%
Thermo Scientific Chemicals

2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 97%

CAS: 28485-18-9 | C4H3ClN2O4S | 210.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54410.06
also known as H54410-06
Price (USD)
503.65
Special offer
Online exclusive
Ends: 30-Jun-2026
592.00
Save 88.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
503.65
Special offer
Online exclusive
Ends: 30-Jun-2026
592.00
Save 88.35 (15%)
Each
Specifications
CAS28485-18-9
Chemical Name or Material2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight209.95
Health Hazard 1H314-H335
View more
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only