Specifications
CAS33763-20-1
Chemical Name or Material4-Methyl-2-phenylthiazole-5-carboxylic acid
Formula Weight219.26
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more