3-Amino-2-methoxypyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-2-methoxypyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H54570.03
also known as H54570-03
1 g
Catalog number H54570.03
also known as H54570-03
Price (USD)
252.65
Online Exclusive
281.00
Save 28.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
252.65
Online Exclusive
281.00
Save 28.35 (10%)
Each
Specifications
CAS893440-50-1
Chemical Name or Material3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
Formula Weight250.1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents, air, heat. Refrigerated (2-4°C).
RUO – Research Use Only