1,3-Cycloheptanedione, 97%
1,3-Cycloheptanedione, 97%
1,3-Cycloheptanedione, 97%
Thermo Scientific Chemicals

1,3-Cycloheptanedione, 97%

CAS: 1194-18-9 | C7H10O2 | 126.155 g/mol
Have Questions?
Catalog NumberQuantity
H54588.03
also known as H54588-03
1 g
Catalog number H54588.03
also known as H54588-03
Price (USD)
171.65
Online Exclusive
191.00
Save 19.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
171.65
Online Exclusive
191.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS1194-18-9
IUPAC Namecycloheptane-1,3-dione
Molecular FormulaC7H10O2
InChI KeyDBOVMTXPZWVYAQ-UHFFFAOYSA-N
SMILESO=C1CCCCC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
1,3-Cyclopentanedione was used in the synthesis of chemical probes for selective labeling of sulfuric acid proteins. is a versatile reagent for synthesis of perhydroazulenes, prostaglandin products (biologically active unsaturated 20-carbon fatty acids including a 5-carbon ring, derived enzymatically ), and Knoevenagel products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclopentanedione was used in the synthesis of chemical probes for selective labeling of sulfuric acid proteins. is a versatile reagent for synthesis of perhydroazulenes, prostaglandin products (biologically active unsaturated 2
RUO – Research Use Only

General References:

  1. Amos B. SmithIII.; Bruce D. Dorsey.; Masashi Ohba.; Andrew T. LupoJr.; Michael S. Malamas. Preparation, reactivity, and spectral properties of 1,3-dioxin vinylogous esters: versatile .beta.-ketovinyl cation equivalents. J. Org. Chem. 1988, 53 (18),4314-4325.
  2. Hideaki Watanabe , Mitsuhiro Iwamoto.; Masahisa Nakada. Preparation of New Chiral Building Blocks: Highly Enantioselective Reduction of Prochiral 1,3-Cycloalkanediones Possessing a Methyl Group and a Protected Hydroxymethyl Group at Their C2 Position with Baker's Yeast or CBS Catalyst.J. Org. Chem. 2005, 70 (12),4652-4658.