1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%
Thermo Scientific Chemicals

1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%

CAS: 762240-99-3 | C4H4ClF3N2O2 | 204.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H54647.03
also known as H54647-03
Price (USD)
118.65
Online Exclusive
132.00
Save 13.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
118.65
Online Exclusive
132.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS762240-99-3
IUPAC Name2-chloro-N'-(2,2,2-trifluoroacetyl)acetohydrazide
Molecular FormulaC4H4ClF3N2O2
InChI KeyDYKIVKLXFDNBMY-UHFFFAOYSA-N
SMILESFC(F)(F)C(=O)NNC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormSolid
Purity≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only