3,4-Dimethoxyphenylmagnesium bromide, 0.5M in THF
3,4-Dimethoxyphenylmagnesium bromide, 0.5M in THF
3,4-Dimethoxyphenylmagnesium bromide, 0.5M in THF
Thermo Scientific Chemicals

3,4-Dimethoxyphenylmagnesium bromide, 0.5M in THF

CAS: 89980-69-8 | C8H9BrMgO2 | 241.37 g/mol
Have Questions?
Catalog NumberQuantity
H54654.AE
also known as H54654-AE
100 mL
Catalog number H54654.AE
also known as H54654-AE
Price (USD)
263.65
Special offer
Online exclusive
Ends: 30-Jun-2026
310.00
Save 46.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
263.65
Special offer
Online exclusive
Ends: 30-Jun-2026
310.00
Save 46.35 (15%)
Each
Specifications
CAS89980-69-8
Chemical Name or Material3,4-Dimethoxyphenylmagnesium bromide
Concentration or Composition (by Analyte or Components)0.5M in THF
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Formula Weight241.37
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Air, oxidizing agents, heat are incompatible. Store under cool place.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. E. J. Corey.; Raman K. Bakshi. ; Saizo Shibata.; Chung Pin Chen.; Vinod K. Singh. A stable and easily prepared catalyst for the enantioselective reduction of ketones. Applications to multistep syntheses. J. Am. Chem. Soc. 1987, 109 (25),7925-7926.
  2. Margaretha Van der Mey.; Armin Hatzelmann.; Ivonne J. Van der Laan.; Geert J. Sterk.; Ulrich Thibaut.; Hendrik Timmerman. Novel Selective PDE4 Inhibitors. 1. Synthesis, Structure-Activity Relationships, and Molecular Modeling of 4-(3,4-Dimethoxyphenyl)-2H-phthalazin-1-ones and Analogues. J. Med. Chem. 2001, 44 (16),2511-2522.