Methyl 4-chlorocarbonylbenzoate, 98%
Methyl 4-chlorocarbonylbenzoate, 98%
Methyl 4-chlorocarbonylbenzoate, 98%
Methyl 4-chlorocarbonylbenzoate, 98%
Thermo Scientific Chemicals

Methyl 4-chlorocarbonylbenzoate, 98%

CAS: 7377-26-6 | C9H7ClO3 | 198.602 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54688.03
also known as H54688-03
Price (USD)
56.65
Online Exclusive
63.00
Save 6.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
63.00
Save 6.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7377-26-6
IUPAC Namemethyl 4-(carbonochloridoyl)benzoate
Molecular FormulaC9H7ClO3
InChI KeyCVXXHXPNTZBZEL-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.0%
4-(Chlorocarbonyl)benzoic Acid Methyl Ester, is a building block, that can be used for the synthesis of various pharmaceutical compounds. It can be used for the preparation of benzophenone residues and also for the synthesis of novel novel bulky hydrophobic retinoids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Chlorocarbonyl)benzoic Acid Methyl Ester, is a building block, that can be used for the synthesis of various pharmaceutical compounds. It can be used for the preparation of benzophenone residues and also for the synthesis of novel novel bulky hydrophobic retinoids.

Solubility
Hydrolyzes in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids. Hygroscopic.
RUO – Research Use Only

General References:

  1. Qiang Tan.; Elizabeth T Birzin.; Wanda Chan.; Yi Tien Yang.; Lee-Yuh Pai.; Edward C Hayes.; Carolyn A DaSilva.; Frank DiNinno.; Susan P Rohrer.; James M Schaeffer.; Milton L Hammond. Estrogen receptor ligands. Part 5: The SAR of dihydrobenzoxathiins containing modified basic side chains Bioorg. Med. Chem. Lett. 2004, 14 (14),3747-3751.
  2. Ramzi F. Sweis.; Julianne A. Hunt.; Florida Kallashi.; Milton L. Hammond.; Ying Chen.; Suzanne S. Eveland.; Qiu Guo.; Sheryl A. Hylandb.; Denise P. Milot.; Anne-Marie Cumiskey.; Melanie Latham.; Raymond Rosa.; Larry Peterson.; Carl P. Sparrow.; Samuel D. Wright.; Matt S. Anderson.; Peter J. Sinclair. 2-(4-Carbonylphenyl)benzoxazole inhibitors of CETP: Scaffold design and advancement in HDLc-raising efficacy. Bioorg. Med. Chem. Lett. 2011, 21 (6),1890-1895.