2-Amino-3-methylpyridine-5-boronic acid pinacol ester, 96%
2-Amino-3-methylpyridine-5-boronic acid pinacol ester, 96%
2-Amino-3-methylpyridine-5-boronic acid pinacol ester, 96%
Thermo Scientific Chemicals

2-Amino-3-methylpyridine-5-boronic acid pinacol ester, 96%

CAS: 1111637-91-2 | C12H19BN2O2 | 234.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H54773.03
also known as H54773-03
Price (USD)
274.65
Special offer
Online exclusive
Ends: 30-Jun-2026
323.00
Save 48.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
274.65
Special offer
Online exclusive
Ends: 30-Jun-2026
323.00
Save 48.35 (15%)
Each
Specifications
CAS1111637-91-2
Chemical Name or Material2-Amino-3-methylpyridine-5-boronic acid pinacol ester
Formula Weight234.1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is used as primary and secondary pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Refrigerated (2-4°C). Incompatible with oxidizing agents.
RUO – Research Use Only