1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%
1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%
1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%
1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%
Thermo Scientific Chemicals

1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%

CAS: 288148-34-5 | C4H5ClN2O2S | 180.606 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H54785.03
also known as H54785-03
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS288148-34-5
IUPAC Name1-methyl-1H-pyrazole-4-sulfonyl chloride
Molecular FormulaC4H5ClN2O2S
InChI KeyRDAVKKQKMLINOH-UHFFFAOYSA-N
SMILESCN1C=C(C=N1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only