1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole, 99+%
1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole, 99+%
1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole, 99+%
Thermo Scientific Chemicals

1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole, 99+%

CAS: 74257-00-4 | C11H12N4O4S | 296.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54792.03
also known as H54792-03
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS74257-00-4
IUPAC Name3-nitro-1-(2,4,6-trimethylbenzenesulfonyl)-1H-1,2,4-triazole
Molecular FormulaC11H12N4O4S
InChI KeySFYDWLYPIXHPML-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>99.0%
1-(2-Mesitylenesulfonyl)-3-Nitro-1H-1,2,4-Triazole is an efficient coupling reagent in the synthesis of oligonucleotides via phosphotriester approach.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Mesitylenesulfonyl)-3-Nitro-1H-1,2,4-Triazole is an efficient coupling reagent in the synthesis of oligonucleotides via phosphotriester approach.

Solubility
Soluble in Chloroform.

Notes
Store at 4°C. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Jörg Rademann.; Morten Grøtli.; Morten Meldal.; Klaus Bock. SPOCC:  A Resin for Solid-Phase Organic Chemistry and Enzymatic Reactions on Solid Phase. zJ. Am. Chem. Soc. 1999, 121 (23),5459-5466.
  2. Alexandra Papanikos.; Jörg Rademann.; Morten Meldal. α-Ketocarbonyl Peptides:  A General Approach to Reactive Resin-Bound Intermediates in the Synthesis of Peptide Isosteres for Protease Inhibitor Screening on Solid Support.J. Am. Chem. Soc. 2001, 123 (10),2176-2181.