1-Amino-4-cyclopentylpiperazine, 97%
1-Amino-4-cyclopentylpiperazine, 97%
1-Amino-4-cyclopentylpiperazine, 97%
Thermo Scientific Chemicals

1-Amino-4-cyclopentylpiperazine, 97%

CAS: 61379-64-4 | C9H19N3 | 169.272 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54831.03
also known as H54831-03
Price (USD)
218.65
Online Exclusive
243.00
Save 24.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
218.65
Online Exclusive
243.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS61379-64-4
IUPAC Name4-cyclopentylpiperazin-1-amine
Molecular FormulaC9H19N3
InChI KeyQYHRIASMJNLWHJ-UHFFFAOYSA-N
SMILESNN1CCN(CC1)C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only