4-(Dimethylamino)pyridine tribromide, 97%
4-(Dimethylamino)pyridine tribromide, 97%
4-(Dimethylamino)pyridine tribromide, 97%
Thermo Scientific Chemicals

4-(Dimethylamino)pyridine tribromide, 97%

CAS: 92976-81-3 | C7H10Br3N2- | 361.883 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55010.14
also known as H55010-14
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Specifications
CAS92976-81-3
Chemical Name or Material4-(Dimethylamino)pyridine tribromide
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight362.89
Health Hazard 1H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only