4-Amino-2-methylpyridine, 98%
4-Amino-2-methylpyridine, 98%
4-Amino-2-methylpyridine, 98%
4-Amino-2-methylpyridine, 98%
Thermo Scientific Chemicals

4-Amino-2-methylpyridine, 98%

CAS: 18437-58-6 | C6H8N2 | 108.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H55036.MD
also known as H55036-MD
Price (USD)
47.65
Online Exclusive
52.70
Save 5.05 (10%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
47.65
Online Exclusive
52.70
Save 5.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS18437-58-6
IUPAC Name2-methylpyridin-4-amine
Molecular FormulaC6H8N2
InChI KeyGNCLPNMQEGMNTG-UHFFFAOYSA-N
SMILESCC1=NC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Stefan Louw.; Alberto S. Pereira.; Frédéric Lynen.; Melissa Hanna-Brown.; Pat Sandra. Serial coupling of reversed-phase and hydrophilic interaction liquid chromatography to broaden the elution window for the analysis of pharmaceutical compounds. Journal of Chromatography A. 2008, 1208 (1-2), 90-94.
  2. Gerd Vanhoenacker.; Emmie Dumont.; Frank David.; Andrew Baker.; Pat Sandra. Determination of arylamines and aminopyridines in pharmaceutical products using in-situ derivatization and liquid chromatography-mass spectrometry. Journal of Chromatography A. 2009, 1216 (16), 3563-3570.