3-(Boc-amino)phenol, 97%
3-(Boc-amino)phenol, 97%
3-(Boc-amino)phenol, 97%
Thermo Scientific Chemicals

3-(Boc-amino)phenol, 97%

CAS: 19962-06-2 | C11H15NO3 | 209.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55059.06
also known as H55059-06
Price (USD)
59.65
Special offer
Online exclusive
Ends: 30-Jun-2026
69.80
Save 10.15 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
59.65
Special offer
Online exclusive
Ends: 30-Jun-2026
69.80
Save 10.15 (15%)
Each
Chemical Identifiers
CAS19962-06-2
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only