4-[(Trimethylsilyl)ethynyl]benzaldehyde, 97%
4-[(Trimethylsilyl)ethynyl]benzaldehyde, 97%
4-[(Trimethylsilyl)ethynyl]benzaldehyde, 97%
Thermo Scientific Chemicals

4-[(Trimethylsilyl)ethynyl]benzaldehyde, 97%

CAS: 77123-57-0 | C12H14OSi | 202.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H55060.03
also known as H55060-03
Price (USD)
57.50
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
57.50
Each
Chemical Identifiers
CAS77123-57-0
IUPAC Name4-[2-(trimethylsilyl)ethynyl]benzaldehyde
Molecular FormulaC12H14OSi
InChI KeyUZQDUXAJFTWMDT-UHFFFAOYSA-N
SMILESC[Si](C)(C)C#CC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store at 4°C. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. William B. Austin.; Norman Bilow.; William J. Kelleghan.; Kreisler S. Y. Lau. Facile synthesis of ethynylated benzoic acid derivatives and aromatic compounds via ethynyltrimethylsilane. J. Org. Chem. 1981, 46 (11), 2280-2286.
  2. Aidan R. McDonald.; Nicole Franssen.; Gerard P.M. van Klink.; Gerard van Koten. Click’ silica immobilisation of metallo-porphyrin complexes and their application in epoxidation catalysis.J. Organomet. Chem. 2009, 694 (14),2153-2162.