4-n-Propoxyaniline, 97%
4-n-Propoxyaniline, 97%
4-n-Propoxyaniline, 97%
4-n-Propoxyaniline, 97%
Thermo Scientific Chemicals

4-n-Propoxyaniline, 97%

CAS: 4469-80-1 | C9H13NO | 151.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55096.06
also known as H55096-06
Price (USD)
185.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
185.00
Each
Add to cart
Specifications
CAS4469-80-1
Chemical Name or Material4-n-Propoxyaniline
ColorBrown
EINECS Number224-740-0
Flash Point110°C (230°F)
View more
4-n-Propoxyaniline is used as a reactant for preparation of aggrecanase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Propoxyaniline is used as a reactant for preparation of aggrecanase inhibitors.

Solubility
Slightly soluble (1.3 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
A. J. de Koning.; Shirley Milkovitch. The Synthesis of a Number of Ethoxyquin Analogues and their Evaluation as Antioxidants in Fish Oil. Lipid / Fett. 1991, 33 (10), 378-382,