3-[(Trimethylsilyl)ethynyl]pyridine, 97%
3-[(Trimethylsilyl)ethynyl]pyridine, 97%
3-[(Trimethylsilyl)ethynyl]pyridine, 97%
Thermo Scientific Chemicals

3-[(Trimethylsilyl)ethynyl]pyridine, 97%

CAS: 80673-00-3 | C10H13NSi | 175.306 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H55101.14
also known as H55101-14
Price (USD)
309.65
Online Exclusive
344.00
Save 34.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
309.65
Online Exclusive
344.00
Save 34.35 (10%)
Each
Specifications
CAS80673-00-3
Chemical Name or Material3-[(Trimethylsilyl)ethynyl]pyridine
ColorColorless to Yellow
Density0.922 g/mL
Flash Point94°C (201°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only