N-Benzyloxycarbonyl-D-glutamic acid 1-benzyl ester, is used as a reagent in the synthesis of acyl peptides that act as immunostimulants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzyloxycarbonyl-D-glutamic acid 1-benzyl ester, is used as a reagent in the synthesis of acyl peptides that act as immunostimulants.
Solubility
Soluble in methanol and acetic acid.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only