(S)-4-Cyano-4'-(2-methylbutyl)biphenyl, 97%
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl, 97%
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl, 97%
Thermo Scientific Chemicals

(S)-4-Cyano-4'-(2-methylbutyl)biphenyl, 97%

CAS: 63799-11-1 | C18H19N | 249.357 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55183.03
also known as H55183-03
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS63799-11-1
IUPAC Name4'-[(2S)-2-methylbutyl]-[1,1'-biphenyl]-4-carbonitrile
Molecular FormulaC18H19N
InChI KeyDNJQGRFZQMOYGM-AWEZNQCLSA-N
SMILESCC[C@H](C)CC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only