4-Cyano-4'-n-octyloxybiphenyl, 97%
4-Cyano-4'-n-octyloxybiphenyl, 97%
4-Cyano-4'-n-octyloxybiphenyl, 97%
Thermo Scientific Chemicals

4-Cyano-4'-n-octyloxybiphenyl, 97%

CAS: 52364-73-5 | C21H25NO | 307.437 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55191.03
also known as H55191-03
Price (USD)
62.65
Online exclusive
69.90
Save 7.25
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
62.65
Online exclusive
69.90
Save 7.25
Each
Chemical Identifiers
CAS52364-73-5
IUPAC Name4'-(octyloxy)-[1,1'-biphenyl]-4-carbonitrile
Molecular FormulaC21H25NO
InChI KeyGPGGNNIMKOVSAG-UHFFFAOYSA-N
SMILESCCCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only