N-Methyl-3-nitroaniline, 97%
N-Methyl-3-nitroaniline, 97%
N-Methyl-3-nitroaniline, 97%
Thermo Scientific Chemicals

N-Methyl-3-nitroaniline, 97%

CAS: 619-26-1 | C7H8N2O2 | 152.153 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H55196.MD
also known as H55196-MD
Price (USD)
59.40
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
59.40
Each
Specifications
CAS619-26-1
Chemical Name or MaterialN-Methyl-3-nitroaniline
ColorWhite to Brown
Formula Weight152.15
Health Hazard 1H302-H317
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only