4-(1-Piperazinyl)aniline, 95%
4-(1-Piperazinyl)aniline, 95%
4-(1-Piperazinyl)aniline, 95%
4-(1-Piperazinyl)aniline, 95%
Thermo Scientific Chemicals

4-(1-Piperazinyl)aniline, 95%

CAS: 67455-41-8 | C10H15N3 | 177.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55224.03
also known as H55224-03
Price (USD)
84.50
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
84.50
Each
Add to cart
Chemical Identifiers
CAS67455-41-8
IUPAC Name4-(piperazin-1-yl)aniline
Molecular FormulaC10H15N3
InChI KeyVAVOYRCCWLRTMS-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)N1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥94.0%
FormSolid
4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Reactive with oxidizing agents. Keep at room temperature.
RUO – Research Use Only

General References:

  1. Hiroshi Koga.; Akira Itoh.; Satoshi Murayama.; Seigo Suzue.; Tsutomu Irikura. Structure-activity relationships of antibacterial 6,7- and 7,8-disubstituted 1-alkyl-1,4-dihydro-4-oxoquinoline-3-carboxylic acids. J. Med. Chem. 1980, 23 (12),1358-1363.
  2. Yasuo Oshiro.; Seiji Sato.; Nobuyuki Kurahashi.; Tatsuyoshi Tanaka.; Tetsuro Kikuchi.; Katsura Tottori.; Yasufumi Uwahodo.; Takao Nishi. Novel Antipsychotic Agents with Dopamine Autoreceptor Agonist Properties: Synthesis and Pharmacology of 7-[4-(4-Phenyl-1-piperazinyl)butoxy]-3,4-dihydro-2(1H)-quinolinone Derivatives.J. Med. Chem. 1998, 41 (5),658-667.