N-Boc-4-bromoaniline, 97%
N-Boc-4-bromoaniline, 97%
N-Boc-4-bromoaniline, 97%
Thermo Scientific Chemicals

N-Boc-4-bromoaniline, 97%

CAS: 131818-17-2 | C11H14BrNO2 | 272.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55226.06
also known as H55226-06
Price (USD)
66.65
Online exclusive
74.20
Save 7.55
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
66.65
Online exclusive
74.20
Save 7.55
Each
Specifications
CAS131818-17-2
Chemical Name or MaterialN-Boc-4-bromoaniline
ColorBrown to White
Formula Weight272.14
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only