4-Acetyl-N-methylaniline, 97%
4-Acetyl-N-methylaniline, 97%
4-Acetyl-N-methylaniline, 97%
Thermo Scientific Chemicals

4-Acetyl-N-methylaniline, 97%

CAS: 17687-47-7 | C9H11NO | 149.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H55301.03
also known as H55301-03
Price (USD)
109.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
109.00
Each
Specifications
CAS17687-47-7
Chemical Name or Material4-Acetyl-N-methylaniline
ColorBrown to White
Formula Weight149.19
Health Hazard 1H334
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only