2-Hydroxy-4-methoxybenzoic acid, 98%
2-Hydroxy-4-methoxybenzoic acid, 98%
2-Hydroxy-4-methoxybenzoic acid, 98%
Thermo Scientific Chemicals

2-Hydroxy-4-methoxybenzoic acid, 98%

CAS: 2237-36-7 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55345.06
also known as H55345-06
Price (USD)
54.65
Online exclusive
61.00
Save 6.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
54.65
Online exclusive
61.00
Save 6.35
Each
Chemical Identifiers
CAS2237-36-7
IUPAC Name2-hydroxy-4-methoxybenzoic acid
Molecular FormulaC8H8O4
InChI KeyMRIXVKKOHPQOFK-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(O)=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only