4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%
4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%
4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%
4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%
Thermo Scientific Chemicals

4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%

CAS: 114772-54-2 | C14H10BrN | 272.145 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55394.14
also known as H55394-14
Price (USD)
191.65
Special offer
Online exclusive
Ends: 30-Jun-2026
225.00
Save 33.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
191.65
Special offer
Online exclusive
Ends: 30-Jun-2026
225.00
Save 33.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS114772-54-2
IUPAC Name4'-(bromomethyl)-[1,1'-biphenyl]-2-carbonitrile
Molecular FormulaC14H10BrN
InChI KeyLFFIEVAMVPCZNA-UHFFFAOYSA-N
SMILESBrCC1=CC=C(C=C1)C1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only