Tri-tert-butyl borate, 97%
Tri-tert-butyl borate, 97%
Tri-tert-butyl borate, 97%
Thermo Scientific Chemicals

Tri-tert-butyl borate, 97%

CAS: 7397-43-5 | C12H27BO3 | 230.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55461.06
also known as H55461-06
Price (USD)
65.65
Online exclusive
72.50
Save 6.85
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
65.65
Online exclusive
72.50
Save 6.85
Each
Specifications
CAS7397-43-5
Chemical Name or MaterialTri-tert-butyl borate
ColorColorless
DOT InformationTransport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density0.811 g/mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only