4,6-Dimethyldibenzothiophene, 97%
4,6-Dimethyldibenzothiophene, 97%
4,6-Dimethyldibenzothiophene, 97%
4,6-Dimethyldibenzothiophene, 97%
Thermo Scientific Chemicals

4,6-Dimethyldibenzothiophene, 97%

CAS: 1207-12-1 | C14H12S | 212.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H55472.MD
also known as H55472-MD
Price (USD)
45.65
Online Exclusive
50.70
Save 5.05 (10%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
50.70
Save 5.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS1207-12-1
IUPAC Name3,12-dimethyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular FormulaC14H12S
InChI KeyKMPJENUWHPZRGZ-UHFFFAOYSA-N
SMILESCC1=CC2=C(SC3=C2C(C)=CC=C3)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)96.0% min
4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Zeng, Y.; Moghadam, P. Z.; Snurr, R. Q. Pore Size Dependence of Adsorption and Separation of Thiophene/Benzene Mixtures in Zeolites. J. Phys. Chem. C 2015, 119 (27), 15263-15273.
  2. Zhang, L.; Fu, W.; Xiang, M.; Wang, W.; He, M.; Tang, T. MgO Nanosheet Assemblies Supported CoMo Catalyst with High Activity in Hydrodesulfurization of Dibenzothiophene. Ind. Eng. Chem. Res. 2015, 54 (21), 5580-5588.