1-Boc-4-(3-hydroxypropyl)piperazine, 97%
1-Boc-4-(3-hydroxypropyl)piperazine, 97%
1-Boc-4-(3-hydroxypropyl)piperazine, 97%
Thermo Scientific Chemicals

1-Boc-4-(3-hydroxypropyl)piperazine, 97%

CAS: 132710-90-8 | C12H24N2O3 | 244.335 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55547.06
also known as H55547-06
Price (USD)
75.65
Online Exclusive
83.40
Save 7.75 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
75.65
Online Exclusive
83.40
Save 7.75 (9%)
Each
Chemical Identifiers
CAS132710-90-8
IUPAC Nametert-butyl 4-(3-hydroxypropyl)piperazine-1-carboxylate
Molecular FormulaC12H24N2O3
InChI KeyLRYRQGKGCIUVON-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCN(CCCO)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)> 96.0%
Appearance (Color)White to pale yellow
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only