Tri-n-butylphenyltin, 97%
Tri-n-butylphenyltin, 97%
Tri-n-butylphenyltin, 97%
Thermo Scientific Chemicals

Tri-n-butylphenyltin, 97%

CAS: 960-16-7 | C18H32Sn | 367.164 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H55596.14
also known as H55596-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS960-16-7
IUPAC Name(2,3,4-tributylphenyl)stannanylidyne
Molecular FormulaC18H29Sn
InChI KeyZMUNITMSSGNSOT-UHFFFAOYSA-N
SMILESCCCCC1=CC=C([Sn])C(CCCC)=C1CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay (GC)≥96.0%
Proton NMRConforms
FormLiquid
It is applied in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in chemical research.

Solubility
Sparingly Soluble (9.9E-5 g/L) (25°C)

Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Dietmar. Seyferth.; Michael A. Weiner. The Preparation of Organolithium Compounds by the Transmetalation Reaction. IV. Some Factors Affecting the Transmetalation Reaction. J. Am. Chem. Soc. 1962, 84 (3),361-364.
  2. Dietmar Seyferth.; Michael Weiner. Communications: Preparation and Some Reactions of Allyllithium. J. Org. Chem. 1959, 24 (9),1395-1396.