2-Nitrofluorene, 98%
2-Nitrofluorene, 98%
2-Nitrofluorene, 98%
Thermo Scientific Chemicals

2-Nitrofluorene, 98%

CAS: 607-57-8 | C13H9NO2 | 211.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55744.06
also known as H55744-06
Price (USD)
39.10
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.10
Each
Chemical Identifiers
CAS607-57-8
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only