2-Amino-4-bromobenzoic acid, 97%
2-Amino-4-bromobenzoic acid, 97%
2-Amino-4-bromobenzoic acid, 97%
Thermo Scientific Chemicals

2-Amino-4-bromobenzoic acid, 97%

CAS: 20776-50-5 | C7H6BrNO2 | 216.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55829.06
also known as H55829-06
Price (USD)
261.65
Special offer
Online exclusive
Ends: 30-Jun-2026
308.00
Save 46.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
261.65
Special offer
Online exclusive
Ends: 30-Jun-2026
308.00
Save 46.35 (15%)
Each
Chemical Identifiers
CAS20776-50-5
IUPAC Name2-amino-4-bromobenzoic acid
Molecular FormulaC7H6BrNO2
InChI KeyBNNICQAVXPXQAH-UHFFFAOYSA-N
SMILESNC1=CC(Br)=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (slightly).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. P Gruhlke.; A Dölle. 1,2,3,4-Tetrahydro-1,4-methanonaphthalenes as model compounds to study the influence of molecular structure on molecular dynamics by 13C NMR relaxation data. J. Chem. Soc., Perkin Trans. 2. 19982 (10), 2159-2164.
  2. Q Liu.; DG Batt.; JS Lippy.; N Surti.; AJ Tebben. Design and synthesis of carbazole carboxamides as promising inhibitors of Bruton's tyrosine kinase (BTK) and Janus kinase 2 (JAK2). J. Chem. Soc., Perkin Trans. 2. 201525 (19), 4265-4269.