(+)-Di-tert-butyl L-tartrate, 99%
(+)-Di-tert-butyl L-tartrate, 99%
(+)-Di-tert-butyl L-tartrate, 99%
Thermo Scientific Chemicals

(+)-Di-tert-butyl L-tartrate, 99%

CAS: 117384-45-9 | C12H22O6 | 262.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H55860.MD
also known as H55860-MD
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS117384-45-9
IUPAC Name1,4-di-tert-butyl 2,3-dihydroxybutanedioate
Molecular FormulaC12H22O6
InChI KeyITWOKJQQGHCDBL-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)C(O)C(O)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point88-92°C
Optical Rotation+9.0° to +13.0° (c=1 in Acetone)
Assay (GC)≥98.5%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only