1-Phenyl-2-trimethylsilylacetylene, 99%
1-Phenyl-2-trimethylsilylacetylene, 99%
1-Phenyl-2-trimethylsilylacetylene, 99%
Thermo Scientific Chemicals

1-Phenyl-2-trimethylsilylacetylene, 99%

CAS: 2170-06-1 | C11H14Si | 174.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H55866.22
also known as H55866-22
Price (USD)
334.65
Online Exclusive
372.00
Save 37.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
334.65
Online Exclusive
372.00
Save 37.35 (10%)
Each
Chemical Identifiers
CAS2170-06-1
IUPAC Nametrimethyl(2-phenylethynyl)silane
Molecular FormulaC11H14Si
InChI KeyUZIXCCMXZQWTPB-UHFFFAOYSA-N
SMILESC[Si](C)(C)C#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤150
Assay (GC)≥98.5%
GC/MS AnalysisConforms
Appearance (Color)Colorless to yellow
FormLiquid
1-Phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized Pt-η-alkyne complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized Pt-η-alkyne complexes.

Solubility
Sparingly Soluble (6.1E-3 g/L) (25°C)

Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ulrik S. Sørensen.; Esteban Pombo-Villar. Copper-free palladium-catalyzed sonogashira-type coupling of aryl halides and 1-aryl-2-(trimethylsilyl)acetylenes. Tetrahedron. 2005, 61 (10),2697-2703.
  2. Alwyn G. Evans.; J. C. Evans.; P. J. Emes.; T. J. Phelan. Reactions of radical anions. Part IX. The radical anion of 1-phenyl-2-trimethylsilylacetylene. J. Chem. Soc. B. 1971, 315-318.