1-Benzoylpiperazine, 97%
1-Benzoylpiperazine, 97%
1-Benzoylpiperazine, 97%
Thermo Scientific Chemicals

1-Benzoylpiperazine, 97%

CAS: 13754-38-6 | C11H14N2O | 190.246 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55896.06
also known as H55896-06
Price (USD)
116.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
116.00
Each
Chemical Identifiers
CAS13754-38-6
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only