2-[(Trimethylsilyl)ethynyl]aniline, 97%
2-[(Trimethylsilyl)ethynyl]aniline, 97%
2-[(Trimethylsilyl)ethynyl]aniline, 97%
2-[(Trimethylsilyl)ethynyl]aniline, 97%
Thermo Scientific Chemicals

2-[(Trimethylsilyl)ethynyl]aniline, 97%

CAS: 103529-16-4 | C11H15NSi | 189.333 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H55930.06
also known as H55930-06
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Add to cart
Specifications
CAS103529-16-4
Chemical Name or Material2-[(Trimethylsilyl)ethynyl]aniline
ColorColorless to Yellow
Density0.94 g/mL
Flash Point110°C (230°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only