2-Acetyl-6-bromopyridine, 97%
2-Acetyl-6-bromopyridine, 97%
2-Acetyl-6-bromopyridine, 97%
Thermo Scientific Chemicals

2-Acetyl-6-bromopyridine, 97%

CAS: 49669-13-8 | C7H6BrNO | 200.035 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55981.03
also known as H55981-03
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.20
Save 8.55 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.20
Save 8.55 (14%)
Each
Chemical Identifiers
CAS49669-13-8
IUPAC Name1-(6-bromopyridin-2-yl)ethan-1-one
Molecular FormulaC7H6BrNO
InChI KeyRUJTWTUYVOEEFW-UHFFFAOYSA-N
SMILESCC(=O)C1=NC(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. EC Constable.; J Lewis. The preparation and coordination chemistry of 2, 2': 6', 2 ″-terpyridine macrocycles—1. Polyhedron.19821 (3), 303-306.
  2. D Lötscher.; S Rupprecht.; H Stoeckli-Evans. Enantioselective catalytic cyclopropanation of styrenes by copper complexes with chiral pinene-[5, 6]-bipyridine ligands. Tetrahedron: Asymmetry.200011 (21), 4341-4357.