Phenyl methanesulfonate, 98%
Phenyl methanesulfonate, 98%
Phenyl methanesulfonate, 98%
Thermo Scientific Chemicals

Phenyl methanesulfonate, 98%

CAS: 16156-59-5 | C7H8O3S | 172.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55995.06
also known as H55995-06
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.50
Save 8.85 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.50
Save 8.85 (15%)
Each
Specifications
CAS16156-59-5
Chemical Name or MaterialPhenyl methanesulfonate
ColorBrown
Formula Weight172.2
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only