Iodine trichloride, 97%
Iodine trichloride, 97%
Iodine trichloride, 97%
Thermo Scientific Chemicals

Iodine trichloride, 97%

CAS: 865-44-1 | Cl3I | 233.254 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H56005.06
also known as H56005-06
Price (USD)
40.80
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.80
Each
Chemical Identifiers
CAS865-44-1
IUPAC Nametrichloro-λ³-iodane
Molecular FormulaCl3I
InChI KeyPAWIVBWALDNUJP-UHFFFAOYSA-N
SMILESCl[I](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Redox Titration)96.0% min
Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.

Solubility
Soluble with decomposition.

Notes
Light Sensitive. Store at -20°C.
RUO – Research Use Only

General References:

  1. John Cyril Evans.; Grace Y. S. Lo. Chlorine nuclear quadrupole coupling in iodine trichloride and in its adduct with aluminum trichloride. Inorg. Chem. 1967, 6 (4),836-837.
  2. R. H. Lamoreaux.; W. F. Giauque. Thermodynamics of iodine trichloride. Entropy and heat capacity from 15 to 325.deg.K. Composition of the equilibrium iodine monochloride and chlorine gas phase. J. Phys. Chem. 1969, 73 (4), 755-762.