2-Amino-3,5-dibromobenzaldehyde, 97%
2-Amino-3,5-dibromobenzaldehyde, 97%
2-Amino-3,5-dibromobenzaldehyde, 97%
2-Amino-3,5-dibromobenzaldehyde, 97%
Thermo Scientific Chemicals

2-Amino-3,5-dibromobenzaldehyde, 97%

CAS: 50910-55-9 | C7H5Br2NO | 278.931 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H56015.06
also known as H56015-06
Price (USD)
65.70
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
65.70
Each
Add to cart
Chemical Identifiers
CAS50910-55-9
IUPAC Name2-amino-3,5-dibromobenzaldehyde
Molecular FormulaC7H5Br2NO
InChI KeyRCPAZWISSAVDEA-UHFFFAOYSA-N
SMILESNC1=C(Br)C=C(Br)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only